missing translation for 'onlineSavingsMsg'
Learn More

Montmorillonite K-10, granules, Thermo Scientific Chemicals

Catalog Number 15387390
Click to view available options
Quantity:
100 g
1 kg
5 kg
This item is not returnable. View return policy
1318-93-0
Al2H2O12Si4
360.307
GUJOJGAPFQRJSV-UHFFFAOYSA-N
montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
71586775
dialuminum;dioxosilane;oxygen(2-);hydrate
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
This item is not returnable. View return policy
1318-93-0
Al2H2O12Si4
360.307
GUJOJGAPFQRJSV-UHFFFAOYSA-N
montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
71586775
dialuminum;dioxosilane;oxygen(2-);hydrate
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 kg
Fieser 15,213
Merck Index 11,6168
Density 0.500 to 0.600 kg/l
Chemical Name or Material Montmorillonite K-10

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'