missing translation for 'onlineSavingsMsg'
Learn More

Morpholine-4-carboxamidine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog Number p-7024776
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
157415-17-3
C5H12BrN3O
210.075
MFCD00051994
IOHSDHMCZXCESZ-UHFFFAOYSA-N
morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide
2806564
morpholine-4-carboximidamide;hydrobromide
C1COCCN1C(=N)N.Br
This item is not returnable. View return policy
157415-17-3
C5H12BrN3O
210.075
MFCD00051994
IOHSDHMCZXCESZ-UHFFFAOYSA-N
morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide
2806564
morpholine-4-carboximidamide;hydrobromide
C1COCCN1C(=N)N.Br

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point ∼155°C to 165°C
Quantity 5 g
Beilstein 7389038
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 210.08
Percent Purity 98%
Chemical Name or Material Morpholine-4-carboxamidine hydrobromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'