missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Adiponectin/Acrp30 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Plate
6 Plates
Unit Size:
1 set
Description
The Mouse Adiponectin/Acrp30 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Adiponectin/Acrp30 in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications
Specifications
| Assay Range | 0.16 - 10ng/mL |
| Assay Sensitivity | 0.007ng/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates |
| Specificity | Natural and Recombinant full-length Mouse Adiponectin. This assay only recognizes full-length Mouse Adiponectin. It does not recognize Mouse gAdiponectin. |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction