missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DMRT2.
Brand: Abnova H00010655-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: TGPRERCTPAGGGAEPRKLSRTPKCARCRNHGVVSCLKGHKRFCRWRDCQCANCLLVVERQRVMAAQVALRRQQATEDKKGLSGKQNNFERKSpecifications
DMRT2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant DMRT2. | |
DMRT2 | |
DMRT2 | |
DMRT2 (NP_006548, 99 a.a. to 190 a.a) partial recombinant protein with GST tag. | |
RUO | |
10655 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006557 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
For Research Use Only