Learn More
Mouse anti-Mouse IgM[a], Clone: DS-1, biotin, Secondary Antibody, BD
Brand: BD Biosciences 553515
Additional Details : Weight : 0.01000kg
Description
The DS-1 antibody reacts specifically with mouse IgM of Igh-C[a] and related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2). It does not react with IgM of Igh-C[b], Igh-C[e], or related hapltypes (e.g., C57BL, SJL, A, AKR, NZB). DS-1 an
Host Species: Mouse
Clone: DS-1
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Mouse NMRI Monoclonal Antibody TC31
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
Mouse anti-Mouse IgM[a] | |
DS-1 | |
Biotin | |
0.5 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.5mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG1 κ |