missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse BID ELISA Kit

Product Code. 17125609
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17125609

Brand: Invitrogen™ EM10RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Mouse BID quantitates mouse BID in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse BID.

BID is a death agonist that heterodimerizes with either agonist BAX or antagonist BCL2. Bid is a member of the BCL-2 family of cell death regulators. BID is a mediator of mitochondrial damage induced by caspase-8 (CASP8); CASP8 cleaves this encoded protein, and the COOH-terminal part translocates to mitochondria where it triggers cytochrome c release. BID is also a pro-apoptotic member of the Bcl 2 family. BID usually exists in an inactive form in the cytosolic fraction of living cells and becomes cleaved and activated by caspase 8 in response to TNF alpha or Fas ligand. Once BID is cleaved, the C-terminal 15 kDa fragment of BID (p15) translocates onto mitochondria and is sufficient to trigger cytochrome c release, resulting in cell apoptosis. BID serves as a direct molecular link between caspase 8 activation and mitochondrial death machinery.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2700049M22Rik; AI875481; apoptic d; apoptic death agonist; apoptotic death agonist BID; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; Bid; BID isoform ES(1 b); BID isoform L(2); BID isoform Si6; desmocollin type 4; FP497; HGNC:1050; Human BID coding sequence; MGC15319; MGC42355; OTTHUMP00000196197; p11 BID; p13 BID; p15 BID; p22 BID
Assay Range 0.41 to 100 ng/mL
Assay Sensitivity 0.41 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 12122
Gene Symbol BID
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target BID
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.