missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL11/Eotaxin DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
1 set
Description
The Mouse CCL11/Eotaxin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CCL11/Eotaxin.
Specifications
Specifications
| Assay Range | 7.81 to 500 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| Gene ID (Entrez) | 6356 |
| Gene Symbol | Ccl11 |
| Host Species | Mouse |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction