missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CD200 (soluble) Fc Recombinant Protein, PeproTech®

Product Code. 30245346
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30245346

Brand: Gibco™ 315411MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

CD200 is a type-1 membrane glycoprotein that contains two immunoglobulin domains, and belongs to the immunoglobulin superfamily. CD200 is a broadly distributed cell surface glycoprotein that interacts with a receptor on myeloid cells (CD200R) and is implicated in locally preventing macrophage activation. Distant, but recognizable, homologues of CD200 have been identified in many herpesviruses and poxviruses. Studies of the related genes in mouse and rat suggest that CD200 may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms of CD200 have been found. Diseases associated with CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O54901
For Use With (Application) ELISA
Formulation Protein with no preservative
Gene ID (Entrez) 17470
Molecular Weight (g/mol) 48.6 kDa
Name CD200
Quantity 1 mg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein
Gene Symbol Cd200
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence QVQVVTQDER EQLYTPASLK CSLQNAQEAL IVTWQKKKAV SPENMVTFSE NHGVVIQPAY KDKINITQLG LQNSTITFWN ITLEDEGCYM CLFNTFGFGK ISGTACLTVY VQPIVSLHYK FSEDHLNITC SATARPAPMV FWKVPRSGIE NSTVTLSHPN GTTSVTSILH IKDPKNQVGK EVICQVLHLG TVTDFKQTVN KGGGPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGK
Activity Measured by its ability to bind immobilized murine sCD200 R1 Fc in a Functional ELISA.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.