missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse CGRP1 ELISA Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
Description
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | P70160 |
| Assay Range | 15.63 to 1000 pg/mL |
| Assay Sensitivity | 9.38 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 12310 |
| Gene Symbol | Calca |
| Interassay CV | <10% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction