missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Flt-3 Ligand (FLT3L), AOF Recombinant Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
Animal Origin Free Reconstitute with 0.1 mg/mL sterile water
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 163 aa.
- Biological Activity: <10 ng/mL
- Molecular Weight: 18.6 kDa
- Animal Origin Free: Yes
Specifications
Specifications
| Accession Number | P49772 |
| For Use With (Application) | Control, ELISA, Functional Assay, Gel Shift, Western Blot |
| Formulation | 10 mM sodium phosphate with no preservative |
| Gene ID (Entrez) | 14256 |
| Molecular Weight (g/mol) | 18.6 kDa |
| Name | Mouse Flt3 Ligand (Flt3L), AOF |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction