missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LRP-4 Protein

Product Code. 18727187
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18727187

Brand: R&D Systems™ 5948LR025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LRP-4 Protein is derived from CHO. The Recombinant Human LRP-4 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAI36669
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 4038
Molecular Weight (g/mol) 191kDa
Name LRP-4
Preparation Method Reconstitute at 300μg/mL in PBS.
Quality Control Testing SDS-PAGE: 180-225kDa, reducing conditions
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human LRP-4 protein, (N-terminus) Human LRP-4 (Ser21-Ser1725) Accession &Num; AAI36669 GGGSGGGSGGGS 6-His (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ser21 & Gly212
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 80%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.