missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IGSF4C/SynCAM4 (NP_694752) VersaClone cDNA

Product Code. 18702328
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18702328 10 μg 10µg
1 options
This item is not returnable. View return policy

Product Code. 18702328

Brand: R&D Systems™ RDC2328

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Mouse IGSF4C/SynCAM4 (NP_694752) VersaClone cDNA is a member of the immunoglobulin superfamily.

The Mouse IGSF4C/SynCAM4 (NP_694752) VersaClone cDNA clone contains the complete ORF for IGSF4C/SynCAM4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IGSF4C/SynCAM4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene mCadm4
Accession Number NP_694752
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 1180bp
pH 8.5
Gene ID (Entrez) 199731
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.