missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-13 Culture Media Quantikine ELISA

Product Code. 18702124
Click to view available options
Quantity:
1 Kit (for 1 Plate)
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18702124

Brand: R&D Systems™ M1300CB1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse IL-13 Culture Media Quantikine ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-13 Culture Media Quantikine ELISA quantifies mouse IL-13 in cell culture supernates (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.80 to 500 pg/mL
Assay Sensitivity 1.5 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Sample Type Cell Culture Supernatant
Specificity Natural and Recombinant Mouse IL-13
Sufficient For 6 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 16163
Quantity 1 Kit (for 1 Plate)
Sample Volume Cell Culture Supernates (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ALRHMGC116789, BHR1interleukin-13, IL13, IL-13 mgC116788, interleukin 13, MGC116786, NC30, P600
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.