missing translation for 'onlineSavingsMsg'
Learn More

Mouse Ki67/MKI67 Antibody, R&D Systems™

Mouse Monoclonal Antibody

Brand:  R&D Systems MAB11612-SP

191.85 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 30279892

  • 248.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ki67/MKI67 Monoclonal antibody specifically detects Ki67/MKI67 in Mouse samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ki67/MKI67
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
4288
Mouse
Purified
Immunohistochemistry, Immunocytochemistry
1088403
Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126
Synthetic Peptide
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.