missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse LIGHT/TNFSF14 DuoSet ELISA

Product Code. 30000595
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000595 1 Kit (for 5 Plates) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000595 Supplier R&D Systems™ Supplier No. DY179405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Mouse LIGHT/TNFSF14 DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant mouse Lymphotoxin-like, exhibits Inducible expression, and competes with HSV Glycoprotein D for HVEM, a receptor expressed by T lymphocytes (LIGHT).

The Mouse LIGHT/TNFSF14 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse LIGHT/TNFSF14.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 250.00 to 16000 pg/mL
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 8740
Gene Symbol Tnfsf14
Kit Contents Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Target LIGHT/TNFSF14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.