missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Neutrophil Elastase/ELA2 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Description
The Mouse Neutrophil Elastase/ELA2 Quantikine ELISA Kit quantifies mouse Neutrophil Elastase/ELA2 in cell culture supernates (50 pg/ml), tissue lysates (13 pg/ml), serum (10 pg/ml), edta plasma (10 pg/ml), heparin plasma (10 pg/ml).
Specifications
Specifications
| Assay Range | 31.30 to 2000 pg/mL |
| Assay Sensitivity | 3.37 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates |
| Specificity | Natural and Recombinant Human GDNF |
| Gene ID (Entrez) | 1991 |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50 μL),Tissue Lysates (13 μL),Serum (10 μL),EDTA Plasma (10 μL),Heparin Plasma (10 μL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction