missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit

Product Code. 18702114
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18702114

Brand: R&D Systems™ MPC900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Proprotein Convertase 9/PCSK9 in cell culture supernates (50 ul), cell lysates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.50 to 4000 pg/mL
Assay Sensitivity 21.9 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Cell Lysates, Cell Lysates, Plasma (EDTA, Heparin), Plasma (EDTA, Heparin), Serum, Serum
Specificity Natural and Recombinant Mouse PCSK9. This kit detects 60 kDa, 53 kDa, and LDLR-complexed Recombinant Mouse PCSK9.
For Use With (Application) ELISA
Gene ID (Entrez) 255738
Gene Symbol Pcsk9
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (10uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1 convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Target Proprotein Convertase 9/PCSK9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.