missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IGF-I
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Insulin-like growth factor 1 (IGF-I) is a polypeptide which belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin
- IGF-I binds to the IGF-I receptor and is a potent activator of the PI3K/AKT pathway; it also activates ERK1/2 signaling
- IGF-I is required for embryonic development, and it is produced mainly in liver in response to a hepatocyte growth hormone
- In the absence of insulin, IGF-I is necessary for the maintenance of human pluripotent stem cells (Wang et al.) Together with interleukin 3 (IL-3), IGF-I stimulates differentiation and proliferation of myeloid cells, and has been shown to regulate lymphopoiesis by stimulating proliferation and differentiation of T and B cells in lymphoid organs (Heemskerk et al.)
Specifications
Specifications
| Name | IBP1, IGF-IA, IGF-IB, IGF1A, Mechano growth factor, MGF, Somatomedin C |
| Quantity | 10 μg |
| Cell Type | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction