missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-13
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
Description
- Interleukin 13 (IL-13) is a type I cytokine, and signals through type I cytokine receptors to activate JAK/STAT and IRS-1/IRS-2 pathways
- IL-13 is produced by T cells and innate lymphoid cells (Pulendran & Artis) and it inhibits production of pro-inflammatory cytokines, prostaglandins, and reactive oxygen and nitrogen species by monocytes and macrophages (Hershey)
- Unlike human B cells, IL-13 has no effect on mouse B cell development and function
- IL-13 promotes eosinophil survival, activation, and recruitment, and also activates mast cells (Hershey)
- IL-13 regulates gastrointestinal parasite expulsion, airway hyperresponsiveness, allergic inflammation, tissue eosinophilia, goblet cell hyperplasia, intracellular parasitism, tissue remodeling, tumor cell growth, and fibrosis
- IL-13 induces hypercontractility of smooth muscles by directly acting on the muscle cells as well as the enteric nerves (Wynn)
Specifications
Specifications
| Name | Interleukin-13, NC30 |
| Quantity | 100 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Monocytes |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction