missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-13
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
Beskrivelse
- Interleukin 13 (IL-13) is a type I cytokine, and signals through type I cytokine receptors to activate JAK/STAT and IRS-1/IRS-2 pathways
- IL-13 is produced by T cells and innate lymphoid cells (Pulendran & Artis) and it inhibits production of pro-inflammatory cytokines, prostaglandins, and reactive oxygen and nitrogen species by monocytes and macrophages (Hershey)
- Unlike human B cells, IL-13 has no effect on mouse B cell development and function
- IL-13 promotes eosinophil survival, activation, and recruitment, and also activates mast cells (Hershey)
- IL-13 regulates gastrointestinal parasite expulsion, airway hyperresponsiveness, allergic inflammation, tissue eosinophilia, goblet cell hyperplasia, intracellular parasitism, tissue remodeling, tumor cell growth, and fibrosis
- IL-13 induces hypercontractility of smooth muscles by directly acting on the muscle cells as well as the enteric nerves (Wynn)
Tekniske data
Tekniske data
| Name | Interleukin-13, NC30 |
| Quantity | 100 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Monocytes |
| Purity or Quality Grade | ≥95% |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion