missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-3
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Interleukin 3 (IL-3) is a species-specific pleiotropic cytokine that promotes the survival and proliferation of pluripotent hematopoietic stem cells and lineage-committed progenitor cells and their differentiation into mature cells of most lineages, including mast cells, basophils, neutrophils, eosinophils, macrophages, dendritic cells, erythrocytes, and megakaryocytes
- IL-3 is produced by activated T cells, and has a physiological role in inflammation and allergies by promoting the secretion of inflammatory mediators such as histamine, IL-4, and IL-6 by mast cells, basophils and eosinophils
- The mouse IL-3 receptor consists of a unique alpha-subunit (CD123) and two beta subunits, one specific for IL-3 (βIL-3), the other shared with the receptors for IL-5 and GM-CSF (beta common chain, βc or CD131)
Specifications
Specifications
| Name | HCGF, Interleukin-3, MCGF, Multi-CSF, P-cell stimulation factor |
| Quantity | 10 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction