missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-5
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Description
- Interleukin 5 (IL-5) is a member of the short-chain 4-α-helical bundle subset of hematopoietic cytokines
- It binds to the specific α receptor, which in turn recruits the common β receptor that initiates signaling via JAK/STAT and MAPK pathways
- IL-5 is produced by Th2 cells, eosinophils, activated mast cells, and ILC2 cells
- IL-5 affects the differentiation, activation, and survival of eosinophils, thus playing an important role in allergic inflammation, asthma, and parasite immunity
- Stimulation of eosinophils with IL-5 leads to their activation, upregulation of CD11b expression, and inhibition of apoptosis (Shearer)
Specifications
Specifications
| Name | B cell differentiation factor II, BCDFII, EDF, Eosinophil differentiation factor, Interleukin-5, TFR |
| Quantity | 100 μg |
| Cell Type | B Cells; Granulocytes and Subsets; Hematopoietic Stem and Progenitor Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction