missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant IL-5

Product Code. p-7217668
Encompass_Preferred
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
This item is not returnable. View return policy

Product Code. 17129441

Brand: STEMCELL Technologies 78049

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 5

  • Interleukin 5 (IL-5) is a member of the short-chain 4-α-helical bundle subset of hematopoietic cytokines
  • It binds to the specific α receptor, which in turn recruits the common β receptor that initiates signaling via JAK/STAT and MAPK pathways
  • IL-5 is produced by Th2 cells, eosinophils, activated mast cells, and ILC2 cells
  • IL-5 affects the differentiation, activation, and survival of eosinophils, thus playing an important role in allergic inflammation, asthma, and parasite immunity
  • Stimulation of eosinophils with IL-5 leads to their activation, upregulation of CD11b expression, and inhibition of apoptosis (Shearer)

Specifications

Name B cell differentiation factor II, BCDFII, EDF, Eosinophil differentiation factor, Interleukin-5, TFR
Quantity 100 μg
Cell Type B Cells; Granulocytes and Subsets; Hematopoietic Stem and Progenitor Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.