missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant Shh
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho & Scott)
- It is expressed during early embryogenesis in a variety of tissues (Finco et al.) During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocyte precursors and neural crest cells and stimulates proliferation, differentiation, and axon growth (Ho & Scott; Martí)
- Shh also regulates T cell differentiation and activation (Crompton et al.) Recombinant Mouse Shh is a fully biologically active molecule
Specifications
Specifications
| Name | HHG-1, HLP3, HPE3, MCOPCB5, SMMCI, Sonic hedgehog, TPT, TPTPS |
| Quantity | 50 μg |
| Cell Type | Neural Stem and Progenitor Cells; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction