missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant TPO
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- Thrombopoietin (TPO) is a key regulator of megakaryocytopoiesis and thrombopoiesis in vitro and in vivo
- TPO stimulates the proliferation and maturation of megakaryocytes and has an important role in regulating the level of circulating platelets in vivo
- TPO also promotes the survival, self-renewal, and expansion of hematopoietic stem cells and primitive multilineage progenitor cells
- It is commonly used with other cytokines such as stem cell factor (SCF) and Flt3/Flk-2 Ligand to promote expansion of primitive hematopoietic cells in culture (Hitchcock & Kaushansky)
- The TPO receptor, c-Mpl, is expressed at all stages of megakaryopoiesis, from hematopoietic stem and progenitor cells to mature platelets
Specifications
Specifications
| Name | c-MPL ligand, Megakaryocyte colony-stimulating factor, MGDF |
| Quantity | 50 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction