Learn More
R&D Systems™ Mouse Stanniocalcin 2/STC-2 Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 9406-SO-050
Description
The Recombinant Mouse Stanniocalcin 2/STC-2 Protein is derived from NS0. The Recombinant Mouse Stanniocalcin 2/STC-2 Protein has been validated for the following applications: Bioactivity.Order Info
Ships at ambient temperature.
Specifications
| O88452 | |
| 8614 | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| <0.10 EU per 1μg of the protein by the LAL method | |
| 75 to 450ng/mL | |
| Unconjugated |
| Lyophilized from a 0.2μ filtered solution in PBS | |
| 31 kDa | |
| Mouse myeloma cell line,NS0-derived mouse Stanniocalcin 2/STC-2 protein Thr25-Arg296,with a C-terminal 6-His tag | |
| RUO | |
| stanniocalcin 2, stanniocalcin-2, STC2, STC-2 | |
| Recombinant protein | |
| Yes |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.