missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse TFF-2 Recombinant Protein, PeproTech®

Product Code. 17812563
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17812563

Brand: Gibco™ 31530100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine TFF-2 is an 11.9 kDa polypeptide of 106 amino acid residues, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing and protection of the gastrointestinal epithelia. These peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain. SP (spasmolytic polypeptide), also designated Trefoil factor 2 (TFF2) precursor, is a trefoil protein that functions to inhibit gastrointestinal motility and gastric acid secretion. SP may also act as a structural component of the gastric mucus, possibly by stabilizing glycoproteins in the mucus gel through interactions with carbohydrate side chains. A downregulation of SP expression is associated with primary gastric cancer, and a progressive loss of this protein is likely to be involved in the early stage of the multi-step gastric carcinogenesis pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03404
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 21785
Molecular Weight (g/mol) 11.9 kDa
Name Mouse TFF-2
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias mSP; pancreatic spasmolytic polypeptide; PSP; SML1; SP; spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Tff2; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1)
Common Name TFF2
Gene Symbol Tff2
Conjugate Unconjugated
Recombinant Recombinant
Sequence EKPSPCRCSR LTPHNRKNCG FPGITSEQCF DLGCCFDSSV AGVPWCFHPL PNQESEQCVM EVSARKNCGY PGISPEDCAS RNCCFSNLIF EVPWCFFPQS VEDCHY
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.