missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TfR2 (NP_056614.3) VersaClone cDNA

Product Code. 18781817
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18781817 10 μg 10µg
1 options
This item is not returnable. View return policy

Product Code. 18781817

Brand: R&D Systems™ RDC3259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene TfR2
Accession Number NP_056614.3
Gene ID (Entrez) 7036
Gene Alias HFE3, HFE3transferrin receptor protein 2, MGC126368, TfR2, TFRC2, transferrin receptor 2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.