missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgG (Total) Mouse ELISA Standard

Product Code. 15518866
Click to view available options
Quantity:
4 pack
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 15518866

Brand: Invitrogen™ 395040065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Component of Mouse IgG total ELISA Ready-SET-Go! Kit
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Quantity 4 pack
Recombinant Recombinant
For Use With (Equipment) Microplate Reader
Former Identifiers eBioscience
Immunoassay Kit Format ELISA Standard
Target Species Primary Mouse
Target Kit Named IgG
Target Species Named Mouse
Target Species Validated Mouse
Target Sub Specificity Total
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.