missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse TRANCE (RANKL) Recombinant Protein

Recombinant Protein

Brand:  Invitrogen™ RP8601

Additional Details : Weight : 0.01000kg

Product Code. 11539121

  • 337.00€ / 10µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Receptor Activator of NF-kB Ligand (RANK Ligand) is a cell-bound marker (CD254) related to the TNF family of proteins. RANK Ligand plays a critical role in bone metabolism,particularly osteoclast differentiation. In addition,RANK Ligand is expressed by some T cells is thought to promote dendritic cell maturation. Recombinant mouse RANK Ligand is a non-glycosylated protein,containing 175 amino acids,with a molecular weight of 19.5 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined in RAW-Blue™ cells and is typically less than 50ng/mL. The specific activity of this protein is 2x10^4 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at-20°C and add 0.1% BSA for additional stability. Amino acid sequence: MPAMMEGSWLD VAQRGKPEAQ PFAHLTINAA SIPSGSHKVT LSSWYHDRGW AKISNMTLSN GKLRVNQDGF YYLYANICFR HHETSGSVPT DYLQLMVYVV KTSIKIPSSH NLMKGGSTKN WSGNSEFHFY SINVGGFFKL RAGEEISIQV SNPSLLDPDQ DATYFGAFKV QDID

Receptor Activator of NF-kB Ligand (RANK Ligand) is a cell-bound marker (CD254) related to the TNF family of proteins. RANK Ligand plays a critical role in bone metabolism, particularly osteoclast differentiation. In addition, RANK Ligand is expressed by some T cells is thought to promote dendritic cell maturation. Recombinant mouse IGF-I is a non-glycosylated protein, containing 70 amino acids, with a molecular weight of 7.6 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of FDC-P1 cells and is typically less than 20 ng/mL. The specific activity of this protein is 5x10^4 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKAA.
Specifications

Specifications

O35235
Control
21943
Mouse CD254 (RANK Ligand)
10μg
-20°C, Avoid Freeze/Thaw Cycles
M-Rank-L
Tnfsf11
Unconjugated
E. coli
Lyophilized
purified
0.1mg/mL
PBS, water with no preservative; pH 7.5
∽19.9
7.5
E. coli
RUO
Mouse CD254 (RANK Ligand)
Mouse
Ms
Recombinant
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions