missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse TSLP Receptor Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0497

Additional Details : Weight : 0.01000kg

Product Code. 15236709

  • 130.00€ / 50µg
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TSLP Receptor Synthetic Peptide, PEP-0497, from Invitrogen.

This peptide corresponds to 21 amino acids near the center of mouse TSLP receptor.PEP-0497 can be used as a blocking peptide with polyclonal antibody PA5-20380.
Specifications

Specifications

57914
purified
Mouse TSLP Receptor
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Mouse
Q8CII9
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Crlf2
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions