Learn More
R&D Systems™ Mouse XL IL-1ra/IL-1F3, Luminex™ Performance Assay
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Mouse |
| Gene ID (Entrez) | 3557 |
| Synonym | DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |