missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g
Unit Size:
100g
10g
1kg
25g
500g
50g
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Features low endotoxin, low level of lipids, fatty acids and hormones, e.g. insulin, T3, T4.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- Free Fatty Acids ≤0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Insulin, T3, T4
Specifications
Specifications
| For Use With (Application) | Mammalian Cell Culture |
| Molecular Weight (g/mol) | 66 |
| Name | CellMaxx™ Bovine Albumin |
| pH Range | 6.5 to 7.5 |
| Purification Method | Chromatography |
| Quantity | 10 g |
| Source | Bovine |
| Storage Requirements | 4°C |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction