missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Collagen Type I from Calf Skin
Suitable for cell adhesion in tissue culture as well as gel formation, platelet aggregation and as a substrate for collagenase
42.29€ - 546.96€
Specifications
For Use With (Application) | Tumor Marker |
---|---|
pH Range | 3.7 |
Source | Calf Skin |
Product Type | Chorionic Gonadotropin from Human Pregnancy Urine |
Content And Storage | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17455293
|
MP Biomedicals™
0215002610 |
10mg |
42.29€
10mg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11406412
|
MP Biomedicals™
0215002680 |
100mg |
261.82€
100mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11416412
|
MP Biomedicals™
0215002683 |
250mg |
546.96€
250mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- CAS Number: 9007-34-5
- EC Number: 232-697-4
- MDL Number: MFCD00130825
- Format: Powder
- Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid
For attachment of primary cultures of epithelioid cells and many other cell types
Specifications
Tumor Marker | |
Calf Skin | |
-20°C |
3.7 | |
Chorionic Gonadotropin from Human Pregnancy Urine | |
Lyophilized |