missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ DOB Medium, Powdered Media

Product Code. p-4474887 Shop All MP Biomedicals Products
Click to view available options
Quantity:
227 g
454 g
1000 g
Unit Size:
1kg
227g
454g
This item is not returnable. View return policy

Product Code. 11357789

Brand: MP Biomedicals™ 114025012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used to create SD or SDA by combining with the appropriate amino acid mixture

  • Use 26.7g powder per liter
  • Contents/liter: 1.7g YNB, 5g Ammonium Sulfate, 20g Dextrose

Yeast cell culture, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae
Quantity 227 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.