Learn More
MP Biomedicals™ Mouse Serum
Used for blocking agents and controls in immunoassays.
Brand: MP Biomedicals™ 08642931
138.82 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.01000kg
Specifications
-20°C | |
Lyophilized | |
10 mL | |
Blocking agents and controls in immunoassays | |
Yes |
Mouse | |
Mouse Serum | |
Filtration | |
5.8 g/dL | |
7.5 |