missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Terrific Broth, For Molecular Biology Dehydrated Media
Shop All MP Biomedicals ProductsSpecifications
Specifications
| Content And Storage | 4°C |
| Form | Dehydrated |
| Media Type | Terrific Broth |
| Product Type | Microbiology Media |
| Formulation | Tryptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.3g/L, Potassium phosphate, Dibasic 12.5g/L |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?