missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MRS (ISO) Agar (DeMan, Rogosa and Sharpe) (Dehydrated)
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50kg
500g
Description
Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ MRS (ISO) Agar (DeMan, Rogosa and Sharpe) (Dehydrated) while adhering to ISO 15214: 1998. This medium is designed to support the growth of all strains of lactobacilli, even the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti.
Use MRS (ISO) Agar for enumeration and isolation of mesophilic lactic acid bacteria from food samples. MRS (ISO) Agar encourages the growth of the lactic acid bacteria, which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus, and Leuconostoc.
MRS (ISO) Agar was designed to conform to the formulation specified in ISO 15214: 1998 Horizontal method for the enumeration of Mesophilic lactic acid bacteria―colony count technique at 30°C.
The medium shows some degree of selectivity, however, if yeasts and mold contamination in the food samples is of concern, these microorganisms can be inhibited by the addition of selective agents like sorbic acid, ciprofloxacin, or clindamycin.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
- Adheres to ISO 15214: 1998 standards.
- Superior performance over tomato juice medium of Briggs1 and the meat extract tomato juice medium of de Man.
MRS (ISO) Agar was designed to conform to the formulation specified in ISO 15214: 1998 Horizontal method for the enumeration of Mesophilic lactic acid bacteria―colony count technique at 30°C.
The medium shows some degree of selectivity, however, if yeasts and mold contamination in the food samples is of concern, these microorganisms can be inhibited by the addition of selective agents like sorbic acid, ciprofloxacin, or clindamycin.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Content And Storage | 10°C to 30°C |
| Description | MRS (ISO) Agar (DeMan, Rogosa and Sharpe) (Dehydrated) |
| Form | Powder |
| Product Type | Agar |
| Quantity | 2.5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction