missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MS 159
Description
MS 159 is a nuclear receptor binding SET structural domain protein 2 (NSD2) Degrader (DC50 = 5.2 μM, Dmax = >82%). Also degrades IKZF3 and IKZF1. Comprises an NSD2 binding moiety joined by a linker to a cereblon E3 ligase ligand. Inhibits proliferation of KMS11 and H929 tumor cells. Bioavailable in vivo.
Specifications
Specifications
| Chemical Name or Material | N-Cyclopropyl-N-(4-((5-(3-(2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetamido)propoxy)pyridin-2-yl)-carbamoyl)benzyl)-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carboxamide |
| CAS | 3031353-59-7 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C43H40N8O10 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?