missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MS 21570

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MS 21570 is a GPR171 antagonist (IC50 = 220 nM). Inhibits BigLEN (Cat.No. 6304) mediated increases in [35S]GTPγS binding in hypothalamic membranes and hyperpolarization of basolateral amygdala pyramidal neurons. Reduces anxiety-like behavior and fear conditioning in mice.
Specifications
Specifications
| Chemical Name or Material | N-Methyl-5-[(phenylmethyl)thio]-1,3,4-thiazadiazol-2-amine |
| CAS | 65373-29-7 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C10H11N3S2 |
| Formula Weight | Observed MW: 237.34 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction