Learn More
Tocris Bioscience™ MS 275
GREENER_CHOICE
HDAC (Class I) inhibitor
Brand:  Tocris Bioscience™ 6208/10
Encompass
Product Code. 15756379
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
MS 275 is a class I HDAC inhibitor (IC50 values are 0.18, 0.74, 44.9 and >100 μM for HDAC1, 3, 8 and 6, respectively). Exhibits antiproliferative effects and induces apoptosis in a range of tumor cell lines in vitro and in vivo. Increases estrogen receptor α- and aromatase expression in breast cancer cells. Inhibits PCB-induced neuronal cell death by preventing HDAC3 binding and histone deacetylation within the synapsin-1 promoter.
Specifications
Specifications
(Pyridin-3-yl)methyl 4-(2-aminophenylcarbamoyl)benzylcarbamate
209783-80-2
>99%