missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MS 436

Product Code. 18747938
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18747938

Brand: Tocris Bioscience™ 5173/50

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Potent and selective BRD4(1) inhibitor

MS 436 is a potent and selective BRD4 bromodomain inhibitor (Ki = 30 - 50 nM for the first bromodomain (BRD4(1)). Exhibits 10-fold selectivity for BRD4(1) over BRD4(2). Blocks BRD4 transcriptional activity in lipopolysaccharide-induced production of both nitric oxide and IL-6 in mouse macrophages (IC50 values are 3.8 and 4.9 μM, respectively). Attenuates melanoma cell proliferation in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 50 mg
Product Type MS 436
Molecular Weight (g/mol) 383.42
Inhibitors MS 436

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.