missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MS 436
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MS 436 is a potent and selective BRD4 bromodomain inhibitor (Ki = 30 - 50 nM for the first bromodomain (BRD4(1)). Exhibits 10-fold selectivity for BRD4(1) over BRD4(2). Blocks BRD4 transcriptional activity in lipopolysaccharide-induced production of both nitric oxide and IL-6 in mouse macrophages (IC50 values are 3.8 and 4.9 μM, respectively). Attenuates melanoma cell proliferation in vitro.
Specifications
Specifications
| Purity | >98% |
| Quantity | 50 mg |
| Product Type | MS 436 |
| Molecular Weight (g/mol) | 383.42 |
| Inhibitors | MS 436 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction