missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MSC 1186
GREENER_CHOICE

Product Code. 18734524
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18734524

Brand: Tocris Bioscience™ 7927/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective SRPK family inhibitor

MSC1186 is a potent and selective inhibitor of serine arginine protein kinases (SRPK) 3, 1 and 2 (IC50 values are 0.6 nM, 3 nM and 81 nM respectively in biochemical assay; EC50 values in NanoBRETassay are 40 nM, 98 nM and 149 nM, respectively). Shows no activity at CLK and DYRK families (IC50 >10 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[[[2-(6-Chloro-7-fluoro-1H-benzimidazol-2-yl)-4-pyrimidinyl]amino]methyl]-2-pyridinyl]-N-methylmethanesulfonamide
CAS 2871698-23-4
Quantity 10 mg
Target SRPK3/STK23
Molecular Formula C19H17ClFN7O2S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.