missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MSC 1186

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MSC1186 is a potent and selective inhibitor of serine arginine protein kinases (SRPK) 3, 1 and 2 (IC50 values are 0.6 nM, 3 nM and 81 nM respectively in biochemical assay; EC50 values in NanoBRETassay are 40 nM, 98 nM and 149 nM, respectively). Shows no activity at CLK and DYRK families (IC50 >10 μM).
Specifications
Specifications
| Chemical Name or Material | N-[3-[[[2-(6-Chloro-7-fluoro-1H-benzimidazol-2-yl)-4-pyrimidinyl]amino]methyl]-2-pyridinyl]-N-methylmethanesulfonamide |
| CAS | 2871698-23-4 |
| Quantity | 10 mg |
| Target | SRPK3/STK23 |
| Molecular Formula | C19H17ClFN7O2S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction