Learn More
Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform
Product Code. p-4525532
Click to view available options
Description:
MSIA D.A.R.T.'S, Custom
MSIA D.A.R.T.'S, Insulin
MSIA D.A.R.T.'S, Protein A
MSIA D.A.R.T.'S, Protein A/G
MSIA D.A.R.T.'S, Protein G
MSIA D.A.R.T.'S, Reloadable Rack
MSIA D.A.R.T.'S, Streptavidin
Format:
Pack of 24 Tips
Pack of 96 Tips
Reload Rack (tips not included)
Unit Size:
Each
Pack of 24
Pack of 96
Product Code. 15182579
Brand: Thermo Scientific™ 991CUS02
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Improve hybrid MS bio-analysis with this exclusive workflow solution for proteins and peptides.
Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform , an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides. Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.
Next generation hybrid Mass Spectrometry bio-analytical workflows:
  • Enables analysis of proteins of interest down to femtomole level
  • Streamlined workflow for protein qualitative & quantative bio-analysis
  • Employs a proprietary molecular trapping micro-column technology in an easy to use pipette tip housing format
  • Drives antibody/antigen binding kinetics and greatly improves assaying of low-abundance protein from a biological sample
Enabling workflows and technologies for biologic and biomarker research:
  • Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
  • Superior specificity and assaying characteristics compared to conventional approaches
  • Allows for hybrid assaying down to femtomole levels
  • Streptavidin and Protein A, G & A/G MSIA D.A.R.T.'S provide end user flexibility to analyze their specific targets of interest
  • Custom MSIA D.A.R.T.'S are available upon request
  • Optimized workflow solution Insulin (endogenous and therapeutic forms)
Requires:
300 μL pipette such as Thermo Scientific™ Versette™ Liquid Handling Platform, Thermo Scientific™ Finnpipette™ Novus i™ Multichannel pipetters (listed in user manual)
Recommended for:
Hybrid mass spectrometry based assays
For research use only
TRUSTED_SUSTAINABILITY
Specifications
Description MSIA D.A.R.T.'S, Custom
Volume (Metric) 300 μL
Storage Requirements 2° to 8°C
Format Pack of 96 Tips
Product Line MSIA D.A.R.T.S