Learn More
Multi-element Ionic Chromatography Cation Standard solution, H2O matrix, Fisher chemical™
Product Code. 12924375 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Product Code. 12924375
Brand: Fisher Chemical CM1773
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Multi-element Ionic Chromatography Cation Standard solution containing 4 components; 10mg/kg each of Lithium (Li+), Sodium (Na+), Potassium (K+), Calcium (Ca2+) for Ion Chromatography H2O matrix
Specifications
Chemical Name or Material Four Components Solution
Packaging Bottle
Analyte or Component Names Lithium, Sodium, Potassium, Calcium
Physical Form Liquid
Concentration 10mg/Kg of each component
Type Multi-component Cationic Standard Solution
Element Li, Na, K, Ca
Solvent or Matrix Water
Quantity 250 mL