Learn More
Shamrock™ Multi-Purpose Blank Tyvek Labels
Provides extra strength, adhesion and permanent marking for your laboratory needs. Shamrock™ Multi-Purpose Blank Tyvek Labels won't discolor, crack or tear, adhere to almost any surface and are available in a range of sizes.
Brand:  Shamrock™ STV2
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11802651
  • 96.04€ / Pack of 1000
Estimated Shipment
09-06-2021
Add to basket
Description
Description
  • Tear-proof material, long-lasting adhesive
  • Does not discolor over time
  • Permanent marking with ballpoint pen, paper pencil and felt marker
  • Labels pre-cut on roll
Specifications
Specifications
Multi-Purpose Blank Tyvek Label
White
20 x 38 mm
20 x 38 mm
Permanent
For Unique and demanding applications
Tyvek