missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Multi-Purpose Blank Tyvek Labels

Product Code. 11892641
Click to view available options
Size (Metric):
20 x 22 mm
20 x 38 mm
38 x 76 mm
51 x 100 mm
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11892641

Brand: Shamrock™ STV1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides extra strength, adhesion and permanent marking for your laboratory needs. Shamrock™ Multi-Purpose Blank Tyvek Labels won't discolor, crack or tear, adhere to almost any surface and are available in a range of sizes.

  • Tear-proof material, long-lasting adhesive
  • Does not discolor over time
  • Permanent marking with ballpoint pen, paper pencil and felt marker
  • Labels pre-cut on roll

Specifications

Adhesive Type Permanent
Color White
Material Tyvek
Product Type Multi-Purpose Blank Tyvek Label
For Use With (Application) For Unique and demanding applications
Size (Metric) 20 x 22 mm
Dimensions (LxW) 20 x 22 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.