Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Accessory, Reagent Bottle
Provides supply containment
Brand:  Agilent BioTek 7140511
Additional Details : Weight : 0.90000kg
Product Code. 16572282
  • 135.00€ / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Autoclavable
  • With cap and connector
Specifications
Specifications
Reagent Bottle
CE and TUV marked. RoHS compliant.
Cap, connector
Autoclavable
For microfill, multiflo fx, precision, bio-tek dispensers and pipetting systems with autoclavable fluid paths